siRNA Details
| Gene Symbol: Cenpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025673.1 | NP_001020844.1 | 6 | 634 | ||
| GenBank mRNA | BC097399.1 | - | 6 | 634 | 7453610 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpu| Gene Symbol: Cenpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025673.1 | NP_001020844.1 | 6 | 634 | ||
| GenBank mRNA | BC097399.1 | - | 6 | 634 | 7453610 | |