siRNA Details
Gene Symbol: Cfap97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014001.1 | NP_001014023.1 | 2 | 824 | ||
XM_006253157.2 | XP_006253219.1 | 3 | 3563 | |||
XM_008771260.1 | XP_008769482.1 | 2 | 1289 | |||
XM_008771261.1 | XP_008769483.1 | 2 | 2571 | |||
XM_008771262.1 | XP_008769484.1 | 2 | 1016 | |||
XM_008771263.1 | XP_008769485.1 | 3 | 817 | |||
XM_008771264.1 | XP_008769486.1 | 1 | 700 | |||
GenBank mRNA | BC082052.1 | - | 2 | 824 | 7132121 |
