siRNA Details
| Gene Symbol: RGD1304810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107314.1 | NP_001100784.1 | 6 | 1959 | ||
| XM_006253222.1 | XP_006253284.1 | 6 | 1791 | |||
| XM_006253223.2 | XP_006253285.1 | 6 | 3533 | |||
| XM_008771300.1 | XP_008769522.1 | 5 | 1890 | |||
| XM_008771301.1 | XP_008769523.1 | 2 | 1919 | |||

RGD1304810
Genomic Map