siRNA Details
| Gene Symbol: Mak16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014002.1 | NP_001014024.1 | 5 | 405 | ||
| GenBank mRNA | BC090035.1 | - | 5 | 405 | 7367321 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mak16| Gene Symbol: Mak16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014002.1 | NP_001014024.1 | 5 | 405 | ||
| GenBank mRNA | BC090035.1 | - | 5 | 405 | 7367321 | |