siRNA Details
| Gene Symbol: Brf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024773.1 | NP_001019944.1 | 3 | 533 | ||
| GenBank mRNA | BC097435.1 | - | 3 | 533 | 7455454 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brf2| Gene Symbol: Brf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024773.1 | NP_001019944.1 | 3 | 533 | ||
| GenBank mRNA | BC097435.1 | - | 3 | 533 | 7455454 | |