siRNA Details
Gene Symbol: LOC100363275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751617.3 | XP_003751665.2 | 19 | 4356 | ||
XM_006253295.2 | XP_006253357.1 | 20 | 4251 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
