siRNA Details
Gene Symbol: Htra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107321.1 | NP_001100791.1 | 3 | 859 | ||
XM_008771355.1 | XP_008769577.1 | 3 | 4002 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Htra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107321.1 | NP_001100791.1 | 3 | 859 | ||
XM_008771355.1 | XP_008769577.1 | 3 | 4002 |