siRNA Details
| Gene Symbol: Ank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107322.2 | NP_001100792.1 | 17 | 1990 | ||
| XM_008771356.1 | XP_008769578.1 | 20 | 2471 | |||
| XM_008771357.1 | XP_008769579.1 | 20 | 2470 | |||
| XM_008771358.1 | XP_008769580.1 | 20 | 3611 | |||
| XM_008771359.1 | XP_008769581.1 | 20 | 2438 | |||

Ank1
Genomic Map