siRNA Details
| Gene Symbol: Slc25a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047880.1 | NP_001041345.1 | 7 | 900 | ||
| GenBank mRNA | AY325139.1 | - | 7 | 900 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a15| Gene Symbol: Slc25a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047880.1 | NP_001041345.1 | 7 | 900 | ||
| GenBank mRNA | AY325139.1 | - | 7 | 900 | ||