siRNA Details
| Gene Symbol: Slc25a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047880.1 | NP_001041345.1 | 4, 5 | 480 | ||
| XM_006253350.2 | XP_006253412.1 | 4, 5 | 916 | |||
| XM_006253351.2 | XP_006253413.1 | 4, 5 | 1307 | |||
| XM_006253352.2 | XP_006253414.1 | 4, 5 | 1133 | |||
| XM_006253353.2 | XP_006253415.1 | 3, 4 | 1961 | |||
| GenBank mRNA | AY325139.1 | - | 4, 5 | 480 | ||

Slc25a15
Genomic Map