siRNA Details
Gene Symbol: Hiatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107334.1 | NP_001100804.1 | 7 | 1116 | ||
GenBank mRNA | BC166784.1 | - | 7 | 1109 | 7935371 | |
FQ230882.1 | - | 7 | 1121 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hiatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107334.1 | NP_001100804.1 | 7 | 1116 | ||
GenBank mRNA | BC166784.1 | - | 7 | 1109 | 7935371 | |
FQ230882.1 | - | 7 | 1121 |