siRNA Details
| Gene Symbol: Nsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107337.1 | NP_001100807.1 | 5 | 1425 | ||
| XM_006253620.2 | XP_006253682.1 | 5 | 1674 | |||
| XM_006253621.2 | XP_006253683.1 | 5 | 1776 | |||
| XM_006253622.2 | XP_006253684.1 | 5 | 1504 | |||
| XM_006253623.2 | XP_006253685.1 | 5 | 1663 | |||
| XM_006253624.2 | XP_006253686.1 | 5 | 1600 | |||
| XM_006253625.2 | XP_006253687.1 | 5 | 1779 | |||
| XM_006253627.2 | XP_006253689.1 | 5 | 1673 | |||
| XM_008771491.1 | XP_008769713.1 | 5 | 1729 | |||
| XM_008771492.1 | XP_008769714.1 | 4 | 640 | |||
| XM_008771493.1 | XP_008769715.1 | 4 | 930 | |||
| XM_008771494.1 | XP_008769716.1 | 5 | 1673 | |||

Nsd1
Genomic Map