siRNA Details
| Gene Symbol: Bicd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033674.1 | NP_001028846.1 | 6, 7 | 2339 | ||
| NM_198765.2 | NP_942060.1 | 6, 7 | 2319 | |||
| GenBank mRNA | AJ250312.1 | - | 6, 7 | 2319 | ||
| AJ250313.1 | - | 3, 4 | 1658 | |||
| BC100660.1 | - | 6, 7 | 2339 | 7935996 | ||

Bicd2
Genomic Map