siRNA Details
Gene Symbol: Susd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107341.1 | NP_001100811.1 | 4 | 534 | ||
XM_006253709.2 | XP_006253771.1 | 4 | 2110 | |||
GenBank mRNA | FQ227785.1 | - | 4 | 719 |
