siRNA Details
| Gene Symbol: Phactr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214457.2 | NP_999622.2 | 8 | 1282 | ||
| GenBank mRNA | AY494977.1 | - | 1151 | |||
| BC098634.1 | - | 8 | 1282 | 7317963 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phactr1| Gene Symbol: Phactr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214457.2 | NP_999622.2 | 8 | 1282 | ||
| GenBank mRNA | AY494977.1 | - | 1151 | |||
| BC098634.1 | - | 8 | 1282 | 7317963 | ||