siRNA Details
| Gene Symbol: Txndc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271330.1 | NP_001258259.1 | 6, 7 | 844 | ||
| GenBank mRNA | FM067602.1 | - | 610 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txndc5| Gene Symbol: Txndc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271330.1 | NP_001258259.1 | 6, 7 | 844 | ||
| GenBank mRNA | FM067602.1 | - | 610 | |||