siRNA Details
| Gene Symbol: Txndc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271330.1 | NP_001258259.1 | 2, 3 | 432 | ||
| GenBank mRNA | FM045749.1 | - | 2, 3 | 437 | ||
| FM067602.1 | - | 2, 3 | 198 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txndc5| Gene Symbol: Txndc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271330.1 | NP_001258259.1 | 2, 3 | 432 | ||
| GenBank mRNA | FM045749.1 | - | 2, 3 | 437 | ||
| FM067602.1 | - | 2, 3 | 198 | |||