siRNA Details
| Gene Symbol: Fam65b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014009.2 | NP_001014031.2 | 3, 4 | 291 | ||
| XM_006253929.2 | XP_006253991.1 | 1, 2 | 789 | |||
| XM_006253930.1 | XP_006253992.1 | 1, 2 | 145 | |||
| XM_008771636.1 | XP_008769858.1 | 1, 2 | 629 | |||
| XM_008771637.1 | XP_008769859.1 | 11 | ||||
| XM_008771638.1 | XP_008769860.1 | 1, 2 | 630 | |||
| XM_008771639.1 | XP_008769861.1 | 1, 2 | 244 | |||
| GenBank mRNA | AY325194.1 | - | 3, 4 | 291 | ||

Fam65b
Genomic Map