siRNA Details
| Gene Symbol: Fam65b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014009.2 | NP_001014031.2 | 3 | 242 | ||
| GenBank mRNA | AY325194.1 | - | 3 | 242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam65b| Gene Symbol: Fam65b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014009.2 | NP_001014031.2 | 3 | 242 | ||
| GenBank mRNA | AY325194.1 | - | 3 | 242 | ||