siRNA Details
| Gene Symbol: Btn1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170337.1 | NP_001163808.1 | 7 | 1460 | ||
| XM_006253970.2 | XP_006254032.1 | 8 | 1839 | |||
| GenBank mRNA | AY327120.1 | - | 7 | 1464 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btn1a1| Gene Symbol: Btn1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170337.1 | NP_001163808.1 | 7 | 1460 | ||
| XM_006253970.2 | XP_006254032.1 | 8 | 1839 | |||
| GenBank mRNA | AY327120.1 | - | 7 | 1464 | ||