siRNA Details
| Gene Symbol: Zfp192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100574.1 | NP_001094044.1 | 5, 6 | 1046 | ||
| XM_008771718.1 | XP_008769940.1 | 5, 6 | 1357 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp192| Gene Symbol: Zfp192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100574.1 | NP_001094044.1 | 5, 6 | 1046 | ||
| XM_008771718.1 | XP_008769940.1 | 5, 6 | 1357 | |||