siRNA Details
| Gene Symbol: Vps41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107355.1 | NP_001100825.1 | 18 | 1531 | ||
| GenBank mRNA | FQ211797.1 | - | 18 | 1526 | ||
| FQ223450.1 | - | 18 | 1534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps41| Gene Symbol: Vps41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107355.1 | NP_001100825.1 | 18 | 1531 | ||
| GenBank mRNA | FQ211797.1 | - | 18 | 1526 | ||
| FQ223450.1 | - | 18 | 1534 | |||