siRNA Details
| Gene Symbol: Zfp438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107358.1 | NP_001100828.1 | 5 | 2248 | ||
| XM_006254021.2 | XP_006254083.1 | 5 | 2484 | |||
| XM_006254023.2 | XP_006254085.1 | 5 | 2246 | |||
| XM_006254027.2 | XP_006254089.1 | 4 | 2094 | |||
| XM_008771729.1 | XP_008769951.1 | 6 | 3687 | |||
| XM_008771730.1 | XP_008769952.1 | 4 | 2710 | |||

Zfp438
Genomic Map