siRNA Details
| Gene Symbol: Mpp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100575.1 | NP_001094045.1 | 9 | 849 | ||
| XM_006254138.2 | XP_006254200.1 | 10 | 2632 | |||
| GenBank mRNA | BC085813.1 | - | 9 | 932 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mpp7| Gene Symbol: Mpp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100575.1 | NP_001094045.1 | 9 | 849 | ||
| XM_006254138.2 | XP_006254200.1 | 10 | 2632 | |||
| GenBank mRNA | BC085813.1 | - | 9 | 932 | ||