siRNA Details
| Gene Symbol: Dip2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107360.1 | NP_001100830.1 | 21 | 2543 | ||
| XM_008771732.1 | XP_008769954.1 | 21 | 2411 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dip2c| Gene Symbol: Dip2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107360.1 | NP_001100830.1 | 21 | 2543 | ||
| XM_008771732.1 | XP_008769954.1 | 21 | 2411 | |||