siRNA Details
| Gene Symbol: Pitrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107363.1 | NP_001100833.1 | 23 | 2657 | ||
| XM_006254186.2 | XP_006254248.1 | 25 | 2944 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pitrm1| Gene Symbol: Pitrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107363.1 | NP_001100833.1 | 23 | 2657 | ||
| XM_006254186.2 | XP_006254248.1 | 25 | 2944 | |||