siRNA Details
| Gene Symbol: Armc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287020.1 | NP_001273949.1 | 6 | 421 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armc3| Gene Symbol: Armc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287020.1 | NP_001273949.1 | 6 | 421 | ||