siRNA Details
| Gene Symbol: Apbb1ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100577.1 | NP_001094047.1 | 12 | 1447 | ||
| GenBank mRNA | BC098839.1 | - | 4 | 408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apbb1ip| Gene Symbol: Apbb1ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100577.1 | NP_001094047.1 | 12 | 1447 | ||
| GenBank mRNA | BC098839.1 | - | 4 | 408 | ||