siRNA Details
| Gene Symbol: Cd226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107370.1 | NP_001100840.1 | 6 | 1046 | ||
| XM_006255025.2 | XP_006255087.1 | 7 | 1861 | |||
| XM_006255028.2 | XP_006255090.1 | 6 | 1497 | |||
| XM_006255029.2 | XP_006255091.1 | 7 | 1581 | |||
| XM_006255031.2 | XP_006255093.1 | 6 | 1217 | |||
| XM_006255032.2 | XP_006255094.1 | 7 | 1486 | |||

Cd226
Genomic Map