siRNA Details
| Gene Symbol: Cd226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107370.1 | NP_001100840.1 | 4 | 730 | ||
| XM_006255025.2 | XP_006255087.1 | 5 | 1545 | |||
| XM_006255028.2 | XP_006255090.1 | 4 | 1181 | |||
| XM_006255029.2 | XP_006255091.1 | 5 | 1265 | |||
| XM_006255031.2 | XP_006255093.1 | 4 | 901 | |||
| XM_006255032.2 | XP_006255094.1 | 5 | 1170 | |||

Cd226
Genomic Map