siRNA Details
| Gene Symbol: Socs6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271149.1 | NP_001258078.1 | 2 | 1297 | ||
| XM_006255033.2 | XP_006255095.1 | 3 | 1650 | |||
| XM_006255034.2 | XP_006255096.1 | 3 | 1534 | |||
| XM_008772132.1 | XP_008770354.1 | 2 | 1545 | |||

Socs6
Genomic Map