siRNA Details
| Gene Symbol: Cndp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007687.1 | NP_001007688.1 | 3 | 376 | ||
| XM_006255009.2 | XP_006255071.1 | 4 | 2098 | |||
| XM_006255010.2 | XP_006255072.1 | 4 | 2099 | |||
| XM_008772135.1 | XP_008770357.1 | 4 | 398 | |||
| GenBank mRNA | BC081877.1 | - | 3 | 376 | 7107931 | |

Cndp1
Genomic Map