siRNA Details
| Gene Symbol: Zfp608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107378.1 | NP_001100848.1 | 5 | 3370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp608| Gene Symbol: Zfp608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107378.1 | NP_001100848.1 | 5 | 3370 | ||