siRNA Details
| Gene Symbol: Afg3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134864.1 | NP_001128336.1 | 16 | 2287 | ||
| GenBank mRNA | BC091419.1 | - | 2 | 653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Afg3l2| Gene Symbol: Afg3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134864.1 | NP_001128336.1 | 16 | 2287 | ||
| GenBank mRNA | BC091419.1 | - | 2 | 653 | ||