siRNA Details
| Gene Symbol: Csf1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029901.1 | NP_001025072.1 | 6 | 685 | ||
| XM_006254813.2 | XP_006254875.1 | 3 | 783 | |||
| XM_008772147.1 | XP_008770369.1 | 4 | 810 | |||
| XM_008772148.1 | XP_008770370.1 | 4 | 832 | |||

Csf1r
Genomic Map