siRNA Details
| Gene Symbol: Fchsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107392.1 | NP_001100862.2 | 6 | 848 | ||
| NM_001109881.1 | NP_001103351.1 | 8 | 674 | |||
| XM_006254635.2 | XP_006254697.1 | 8 | 681 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fchsd1| Gene Symbol: Fchsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107392.1 | NP_001100862.2 | 6 | 848 | ||
| NM_001109881.1 | NP_001103351.1 | 8 | 674 | |||
| XM_006254635.2 | XP_006254697.1 | 8 | 681 | |||