siRNA Details
| Gene Symbol: Etf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008344.1 | NP_001008345.1 | 10 | 1271 | ||
| GenBank mRNA | BC085902.1 | - | 10 | 1271 | 7193574 | |
| FQ224770.1 | - | 1 | 12 | |||
| FQ227237.1 | - | 10 | 1300 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etf1| Gene Symbol: Etf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008344.1 | NP_001008345.1 | 10 | 1271 | ||
| GenBank mRNA | BC085902.1 | - | 10 | 1271 | 7193574 | |
| FQ224770.1 | - | 1 | 12 | |||
| FQ227237.1 | - | 10 | 1300 | |||