siRNA Details
| Gene Symbol: Ctnna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007145.1 | NP_001007146.1 | 14 | 1998 | ||
| XM_008772037.1 | XP_008770259.1 | 8 | 906 | |||
| GenBank mRNA | BC085789.1 | - | 14 | 1998 | 7108824 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctnna1| Gene Symbol: Ctnna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007145.1 | NP_001007146.1 | 14 | 1998 | ||
| XM_008772037.1 | XP_008770259.1 | 8 | 906 | |||
| GenBank mRNA | BC085789.1 | - | 14 | 1998 | 7108824 | |