siRNA Details
| Gene Symbol: Elp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034145.1 | NP_001029317.1 | 8 | 746 | ||
| GenBank mRNA | BC100661.1 | - | 8 | 746 | 7935317 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elp2| Gene Symbol: Elp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034145.1 | NP_001029317.1 | 8 | 746 | ||
| GenBank mRNA | BC100661.1 | - | 8 | 746 | 7935317 | |