siRNA Details
Gene Symbol: Zfp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013141.1 | NP_001013159.1 | 3 | 921 | ||
GenBank mRNA | BC089850.1 | - | 3 | 921 | 7375328 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013141.1 | NP_001013159.1 | 3 | 921 | ||
GenBank mRNA | BC089850.1 | - | 3 | 921 | 7375328 |