siRNA Details
| Gene Symbol: Dsc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107402.1 | NP_001100872.1 | 10 | 1693 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dsc3| Gene Symbol: Dsc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107402.1 | NP_001100872.1 | 10 | 1693 | ||