siRNA Details
| Gene Symbol: Cdh16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012055.1 | NP_001012055.1 | 7 | 835 | ||
| GenBank mRNA | BC081996.1 | - | 7 | 835 | 7130691 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh16| Gene Symbol: Cdh16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012055.1 | NP_001012055.1 | 7 | 835 | ||
| GenBank mRNA | BC081996.1 | - | 7 | 835 | 7130691 | |