siRNA Details
| Gene Symbol: Cdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107407.1 | NP_001100877.1 | 9 | 1409 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh5| Gene Symbol: Cdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107407.1 | NP_001100877.1 | 9 | 1409 | ||