siRNA Details
| Gene Symbol: Gpr114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107410.1 | NP_001100880.1 | 9 | 925 | ||
| XM_006255101.2 | XP_006255163.1 | 10 | 998 | |||
| XM_008772297.1 | XP_008770519.1 | 10 | 983 | |||
| XM_008772300.1 | XP_008770522.1 | 9 | 1094 | |||
| XM_008772301.1 | XP_008770523.1 | 10 | 1022 | |||
| XM_008772302.1 | XP_008770524.1 | 10 | 944 | |||
| XM_008772303.1 | XP_008770525.1 | 10 | 976 | |||
| XM_008772304.1 | XP_008770526.1 | 10 | 1283 | |||
| XM_008772305.1 | XP_008770527.1 | 10 | 944 | |||
| XM_008772306.1 | XP_008770528.1 | 10 | 985 | |||
| XM_008772307.1 | XP_008770529.1 | 10 | 982 | |||
| XM_008772308.1 | XP_008770530.1 | 10 | 1017 | |||
| XM_008772309.1 | XP_008770531.1 | 10 | 993 | |||
| XM_008772310.1 | XP_008770532.1 | 10 | 971 | |||
| XM_008772311.1 | XP_008770533.1 | 10 | 936 | |||
More Information
Set Membership: |
|

Gpr114
Genomic Map