siRNA Details
| Gene Symbol: Papd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107416.1 | NP_001100886.1 | 8 | 1096 | ||
| XM_006255261.1 | XP_006255323.1 | 10 | 1706 | |||
| XM_006255263.2 | XP_006255325.1 | 10 | 1706 | |||
| XM_008772403.1 | XP_008770625.1 | 11 | 1717 | |||

Papd5
Genomic Map