siRNA Details
| Gene Symbol: Cenpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024257.1 | NP_001019428.1 | 12 | 1366 | ||
| XM_006255494.2 | XP_006255556.1 | 13 | 1484 | |||
| XM_006255495.2 | XP_006255557.1 | 13 | 1443 | |||
| GenBank mRNA | BC093396.1 | - | 12 | 1366 | 7388929 | |

Cenpt
Genomic Map