siRNA Details
| Gene Symbol: Tango6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255568.2 | XP_006255630.1 | 2 | 262 | ||
| XM_008772562.1 | XP_008770784.1 | 2 | 262 | |||
| XM_226428.8 | XP_226428.6 | 2 | 262 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tango6| Gene Symbol: Tango6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255568.2 | XP_006255630.1 | 2 | 262 | ||
| XM_008772562.1 | XP_008770784.1 | 2 | 262 | |||
| XM_226428.8 | XP_226428.6 | 2 | 262 | |||