siRNA Details
| Gene Symbol: Fuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107429.1 | NP_001100899.1 | 10, 11 | 1016 | ||
| XM_006255624.2 | XP_006255686.2 | 10, 11 | 1028 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fuk| Gene Symbol: Fuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107429.1 | NP_001100899.1 | 10, 11 | 1016 | ||
| XM_006255624.2 | XP_006255686.2 | 10, 11 | 1028 | |||