siRNA Details
| Gene Symbol: Clec18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255608.2 | XP_006255670.1 | 8 | 1647 | ||
| XM_006255609.2 | XP_006255671.1 | 9 | 1533 | |||
| XM_006255610.2 | XP_006255672.1 | 8 | 1647 | |||
| XM_006255611.2 | XP_006255673.1 | 8 | 1647 | |||
| XM_006255612.2 | XP_006255674.1 | 8 | 1647 | |||
| XM_008772568.1 | XP_008770790.1 | 8 | 1647 | |||

Clec18a
Genomic Map