siRNA Details
| Gene Symbol: Sdr42e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271269.1 | NP_001258198.1 | 3 | 641 | ||
| GenBank mRNA | FN798906.1 | - | 2 | 523 | ||
| FQ103187.1 | - | 3 | 642 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdr42e1| Gene Symbol: Sdr42e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271269.1 | NP_001258198.1 | 3 | 641 | ||
| GenBank mRNA | FN798906.1 | - | 2 | 523 | ||
| FQ103187.1 | - | 3 | 642 | |||